missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Monoclonal Antibody (1A8-Ly6g), Brilliant Violet™ 480, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen 414-9668-80
This item is not returnable.
View return policy
Description
Ly6G (Lymphocyte antigen 6 complex locus G6D) is a component of the myeloid differentiation antigen Gr-1, together with Ly6C. The Ly6G protein has a molecular weight of approximately 21-25 kDa and is expressed by myeloid derived cells in the bone marrow. Ly6G is a good marker for detection of peripheral neutrophils, monocytes and granulocytes. Expression of Gr-1 in bone marrow correlates with granulocyte differentiation and maturation. The physiological role of Ly6G remains unclear but studies have shown its involvement in anti-tumor response, and studies with neutrophils suggest that Ly6G ligation may have an inhibitory effect on local immune responses.Specifications
Ly-6G | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P35461 | |
Ly6g | |
Affinity chromatography | |
RUO | |
546644 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
1A8-Ly6g | |
Brilliant Violet 480 | |
Ly6g | |
Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction