missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Monoclonal Antibody (1A8-Ly6g), Brilliant Violet™ 480, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£112.00 - £262.00
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8-Ly6g |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17897021
|
Invitrogen
414-9668-80 |
25 μg |
£112.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17807031
|
Invitrogen
414-9668-82 |
100 μg |
£262.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly6G (Lymphocyte antigen 6 complex locus G6D) is a component of the myeloid differentiation antigen Gr-1, together with Ly6C. The Ly6G protein has a molecular weight of approximately 21-25 kDa and is expressed by myeloid derived cells in the bone marrow. Ly6G is a good marker for detection of peripheral neutrophils, monocytes and granulocytes. Expression of Gr-1 in bone marrow correlates with granulocyte differentiation and maturation. The physiological role of Ly6G remains unclear but studies have shown its involvement in anti-tumor response, and studies with neutrophils suggest that Ly6G ligation may have an inhibitory effect on local immune responses.Specifications
Ly-6G | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G | |
Ly6g | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ly6g |
1A8-Ly6g | |
Flow Cytometry | |
Brilliant Violet 480 | |
Rat | |
Mouse | |
P35461 | |
546644 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title