missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, APC, Clone: 1A8, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £256.00
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972208
|
EnquireBio
QAB56-APC-25ug |
25 μg |
£71.60
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15962208
|
EnquireBio
QAB56-APC-100ug |
100 μg |
£256.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Ly-6G | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
546644 | |
IgG2a κ | |
Protein A/G Purified |
1A8 | |
Flow Cytometry | |
APC | |
RUO | |
XP_006521677 | |
Ly6g | |
Primary | |
Store at 2-4°C. |