missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Lyovapor™ L-200
Freeze dryer with Infinite-Control™
Brand: BUCHI 11L200IN1622000001
12449.58 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- High quality freeze drying (-55°C, 6 kg)
- High level of automation
- Infinite Control™ includes:
- Easy method creation
- Method running
- Data logging
- Chart recording in real-time and interruption if needed
Specifications
6 kg | |
6 | |
Stainless Steel | |
Drying chamber manifold | |
23.15 in. |
13.2 lb. | |
-55°C to +30°C | |
n.a. | |
20.1 in. | |
240 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction