missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Lyovapor™ L-200
Freeze dryer with Infinite-Control™
Brand: BUCHI 11L200IN1622003001
Description
- High quality freeze drying (-55°C, 6 kg)
- High level of automation
- Infinite Control™ includes:
- Easy method creation
- Method running
- Data logging
- Chart recording in real-time and interruption if needed
Specifications
6 kg | |
6 | |
Stainless Steel | |
Vacuum pump, Drying chamber manifold | |
23.15 in. |
13.2 lb. | |
-55°C to +30°C | |
Edwards nXDS6iC | |
20.1 in. | |
240 V |