missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lysing Solution, Whole Blood Lysing Solution
Lysing Solution, Whole Blood Lysing Solution
£219.00 - £474.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Product Type | Whole Blood Lysing Solution |
Shipping Condition | Room Temperature |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10630734
|
Invitrogen™
GAS010 |
25 mL |
£219.00
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10292873
|
Invitrogen™
GAS010S100 |
100 mL |
£474.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
Specifications
Store at room temperature. |
|
Whole Blood Lysing Solution | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.