Learn More
Thermo Scientific™ Lysis Buffer for MagJET Plant RNA Kit
The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction.
Brand: Thermo Scientific™ R2541
Code : 88
Additional Details : Weight : 0.11000kg
Description
The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
6X | |
MagJET Plant gDNA Kit | |
72 mL |
Lysis Buffer | |
MagJET |
For Research Use Only. Not for use in diagnostic procedures.