Learn More
Thermo Scientific™ Lysis Buffer for MagJET Plant RNA Kit
The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction.
£69.00 - £186.00
Specifications
Buffer | Lysis Buffer |
---|---|
For Use With (Equipment) | MagJET Plant gDNA Kit |
Product Line | MagJET |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13535220
|
Thermo Scientific™
R2542 |
140 mL |
£186.00
140mL |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13525220
|
Thermo Scientific™
R2541 |
72 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Lysis Buffer | |
MagJET Plant gDNA Kit | |
MagJET |
For Research Use Only. Not for use in diagnostic procedures.