missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Buffer for MagJET Plant RNA Kit

The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction.

Brand:  Thermo Scientific™ R2542

176.70 GBP valid until 2024-12-31
Use promo code "21969" to get your promotional price.



Code : 88

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 13535220

  • £186.00 / 140mL
Estimated Shipment: 16-09-2024
to see stock.

Description

Description

The Thermo Scientific MagJET Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.

Specifications

Specifications

10X
MagJET Plant gDNA Kit
140 mL
Lysis Buffer
MagJET
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.