missing translation for 'onlineSavingsMsg'
Learn More

Melford Lysozyme from Chicken Egg White

Product Code. 30018448
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 30018448

Brand: Melford L3810010.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.

Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.

Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.

Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.

Specifications

CAS 12650-88-3
Quantity 10 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.