missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Lysozyme from Chicken Egg White
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Description
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.
Specifications
Specifications
| CAS | 12650-88-3 |
| Quantity | 5 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction