missing translation for 'onlineSavingsMsg'
Learn More
m-Cresol, 99%, Thermo Scientific Chemicals Product Code.: 10367523

m-Cresol, 99%, Thermo Scientific Chemicals

Product Code. 10367523
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10367523

Brand: Thermo Scientific Acros 110581000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluolShow MoreShow Less
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Melting Point 8.0°C to 10.0°C
Color Colorless to Red
Density 1.0300g/mL
Boiling Point 203.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OH
Refractive Index 1.5390 to 1.5420
Quantity 100 g
Beilstein 06,373
Merck Index 15,2564
Specific Gravity 1.03
Solubility Information Solubility in water: in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Cresol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468275 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number110585000,110581000,110580010,110580025,110580100
Lot NumberA0454096 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number110585000,110581000,110580010,110580025,110580100
Lot NumberA0465615 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number110585000,11058021,110580010,110581000,110580025,110580100
Lot NumberA0460558 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number110585000,11058021,110580010,110581000,110580025,110580100
Lot NumberA0454096 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number110585000,11058021,110580010,110581000,110580025,110580100
compliance-icons compliance-icons
Product Identifier
  • m-Cresol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
m-Cresol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.