Learn More
m-Phenetidine, 98%, Thermo Scientific Chemicals
£29.30 - £94.10
Chemical Identifiers
CAS | 621-33-0 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.18 |
MDL Number | MFCD00007785 |
InChI Key | WEZAHYDFZNTGKE-UHFFFAOYSA-N |
Synonym | m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine |
PubChem CID | 12120 |
IUPAC Name | 3-ethoxyaniline |
SMILES | CCOC1=CC=CC(N)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10656544
|
Thermo Scientific Acros
186720050 |
5 mL | Glass bottle |
£29.30
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377220
|
Thermo Scientific Acros
186720250 |
25 mL | Glass bottle |
£94.10
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
621-33-0 | |
137.18 | |
WEZAHYDFZNTGKE-UHFFFAOYSA-N | |
12120 | |
CCOC1=CC=CC(N)=C1 |
C8H11NO | |
MFCD00007785 | |
m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine | |
3-ethoxyaniline |
Specifications
621-33-0 | |
100.0 | |
1.0300g/mL | |
>112°C | |
98% | |
C2H5OC6H4NH2 | |
MFCD00007785 | |
m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine | |
CCOC1=CC=CC(N)=C1 | |
137.18 | |
137.18 | |
Liquid |
97.5 | |
Brown | |
248.0°C | |
Authentic | |
C8H11NO | |
1.5650 to 1.5670 | |
1.03 | |
WEZAHYDFZNTGKE-UHFFFAOYSA-N | |
3-ethoxyaniline | |
12120 | |
98% | |
m-Phenetidine, 98% |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-680-2
missing translation for 'rtecsNumber' : SI5400000
missing translation for 'tsca' : TSCA
RUO – Research Use Only