missing translation for 'onlineSavingsMsg'
Learn More
m-Phenylenediamine, 99+%, Thermo Scientific Chemicals Product Code.: 10563101

m-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Product Code. 10563101
25 g, Glass bottle
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10563101

Brand: Thermo Scientific Acros 130560250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-45-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007799
InChI Key WZCQRUWWHSTZEM-UHFFFAOYSA-N
Synonym m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamineShow MoreShow Less
PubChem CID 7935
ChEBI CHEBI:8092
IUPAC Name benzene-1,3-diamine
SMILES C1=CC(=CC(=C1)N)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 65.0°C
Color Brown-Gray or Tan to Yellow
Boiling Point 282.0°C to 284.0°C
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(NH2)2
Quantity 25 g
Beilstein 03, IV, 79
Merck Index 15, 7395
Solubility Information Solubility in water: 350g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 108.14
Percent Purity 99+%
Physical Form Pellets
Chemical Name or Material m-Phenylenediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462878 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number130562500,130565000,130560050,130560250,130560010
Lot NumberA0462068 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number130562500,130565000,130560050,130560250,130560010
Lot NumberA0460706 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number130562500,130565000,130560050,130560250,130560010
Lot NumberA0438819 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number130562500,130565000,130560050,130560250,130560010
Lot NumberA0433246 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number130562500,130565000,130560050,130560250,130560010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
m-Phenylenediamine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.