Learn More
Brady™ M211 and M210 Indoor Outdoor Vinyl Labels
Vinyl labels with high adhesion to difficult surfaces, as well as outdoor durability.
Brand:  Brady™ 139740
33.66 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15971011
  • £39.90 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
B-595 vinyl will adhere to difficult, low surface energy items including PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics. The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough pipes and paper jacketed pipes.
  • Abrasion, cold, fuel and oil resistant
  • Outdoor durable
  • Ideal for general laboratory identification and well plate labeling
Specifications
Specifications
Black on Gray
Vinyl (B-595)
0.25 in.
6.4 mm
Cartridge, Ribbon
Electrical Labeling, Laboratory Labeling, Warehouse Labeling
BMP21-LAB Label Printer, BMP21-PLUS Label Printer, Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer
Thermal Transfer
0.75 in.
19 mm
2011/65/EU and 2015/863