Learn More
Tocris Bioscience™ M8 B hydrochloride
Potent and selective TRPM8 channel blocker
Brand:  Tocris Bioscience™ 5324/50
526.98 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18729967
  • £677.00 / 50mg
Explore more special offers
Description
Description
M8 B hydrochloride is a potent and selective TRPM8 channel blocker. Blocks cold- and icilin -induced TRPM8 activation in vitro (IC50 values are 7.8 and 26.9 nM respectively). Exhibits no effect at other TRP channels (IC50 > 20 μM). Reduces body temperature in vivo.
Specifications
Specifications
>99%
M8 B hydrochloride
M8 B hydrochloride
50 mg
432.96