missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ M8 B hydrochloride

Potent and selective TRPM8 channel blocker

Specifications

Purity >99%
Product Type M8 B hydrochloride
Molecular Weight (g/mol) 432.96
Inhibitors M8 B hydrochloride
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18732689
View Documents
Tocris Bioscience™
5324/10
10 mg N/A
N/A
18729967
View Documents Promotion Details
Tocris Bioscience™
5324/50
50 mg N/A
N/A
Description

Description

Potent and selective TRPM8 channel blocker. Blocks cold- and icilin -induced TRPM8 activation in vitro (IC50 values are 7.8 and 26.9 nM respectively). Exhibits no effect at other TRP channels (IC50 > 20 μM). Reduces body temperature in vivo.
Specifications

Specifications

>99%
M8 B hydrochloride
432.96
M8 B hydrochloride
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only