Learn More
Thermo Scientific™ MAbPac™ HIC-10 HPLC Column
Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.
£210.09 - £1713.53
Specifications
Particle Size | 5 μm |
---|---|
pH | 2 to 8 |
Pore Size | 1000 Å |
Quantity | Each |
Temperature | 60°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
15360411
|
Thermo Scientific™
088480 |
100 mm | 4.6 mm |
£1185.90
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
15380411
|
Thermo Scientific™
088481 |
250 mm | 4.6 mm |
£1713.53
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
15300421
|
Thermo Scientific™
088482 |
10 mm | 4.6 mm |
£210.09
Pack of 2 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
|||||
Description
Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.
Unique Chemistry Designed for Optimal Selectivity of mAbs
The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.
- Excellent biocompatibility and extremely low carryover
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phases
- Rugged column stability
Specifications
5 μm | |
1000 Å | |
60°C | |
Hydrophobic Interaction | |
Responsibly packaged |
2 to 8 | |
Each | |
MAbPac | |
Hydrophobic Interaction |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.