missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns

Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.

Brand:  Thermo Scientific™ 078656

858.84 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12673085

  • £864.30 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.

Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  •  Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5 and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
2 mL/min
5000 psi (345 bar)
2 to 12
Each
Ambient to 60°C
MAbPac
Ion Exchange
PEEK
Bio-LC
4 mm
50 mm
5 μm
Nonporous
50% Acetonitrile
Sulfonate
Spherical, Nonporous Polymer
Ion Exchange
Analytical Column
Responsibly packaged
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns > I.D. x L: 4 x 50mm; Particle Size: 5μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.