Learn More
Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns
BIOPHARMA
Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.
£373.86 - £2803.95
Specifications
pH 2 to 12
Pore Size Nonporous
Temperature Ambient to 60°C
Column Type Ion Exchange
Phase Ion Exchange
View More Specs
Products 14
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Column Format Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Column Format Price Quantity & Availability  
11372063
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
074625
250 mm 4 mm 10 μm Analytical Column
£1185.80
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
11332083
View Documents Promotion Details
Thermo Scientific™
075602
150 mm 4 mm 10 μm Analytical Column
£1020.08
Each
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
11342083
View Documents
Thermo Scientific™
075603
50 mm 4 mm 10 μm Analytical Column
£889.43
Each
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
11391993
View Documents
Thermo Scientific™
075604
250 mm 2 mm 10 μm Analytical Column
£1145.70
Each
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
15540244
View Documents
Thermo Scientific™
088784
250 mm 9 mm 10 μm Preparative Column
£2803.95
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
15124725
View Documents Promotion Details
Thermo Scientific™
085198
150 mm 4 mm 5 μm Analytical Column
£1407.00
Each
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
15134725
View Documents
Thermo Scientific™
082674
50 mm 4.6 mm 5 μm Analytical Column
£1758.75
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
15154725
View Documents
Thermo Scientific™
085209
150 mm 4.6 mm 5 μm Analytical Column
£2110.50
Each
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
15144725
View Documents
Thermo Scientific™
082673
250 mm 4.6 mm 5 μm Analytical Column
£2346.68
Each
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
12663085
View Documents
Thermo Scientific™
077907
50 mm 4 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12683085
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
078655
250 mm 4 mm 5 μm Analytical Column
£1889.40
Each
In Stock
to see stock.
Add to basket
12673085
View Documents
Thermo Scientific™
078656
50 mm 4 mm 5 μm Analytical Column
£864.30
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
11382023
View Documents
Thermo Scientific™
075749
50 mm 2 mm 10 μm Guard Column
£394.97
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
11362063
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
074631
50 mm 4 mm 10 μm Guard Column
£373.86
Each
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  • Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5, and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
Specifications
Specifications
2 to 12
Ambient to 60°C
Ion Exchange
Nonporous
Ion Exchange
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns