Learn More
Cytiva MabSelect Sure™ Chromatography Media
Protein A affinity medium for capturing MAbs at process scale. Cytiva MabSelect SureTM Chromatography Media is used in the commercial manufacture of monoclonal antibodies.
Brand: Cytiva 17-5438-01
Description
Has an alkali-tolerant rProteain A ligand designed to enhance protease resistance to produce monoclonal antibodies- Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
- Design gives enhanced protease resistance resulting in lower ligand leakage
- Generic elution profile for different monoclonal antibodies enables platform approach to purification
- High-flow agarose matrix allows the processing of large feed volumes
- Used in the commercial manufacture of monoclonal antibodies
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
25 mL | |
For Use of Rigorous and Cost-Effective CIP and Sanitization Protocols, Manufacture of Monoclonal Antibodies | |
30 mg Human lgG/mL (at 2.4 min. residence time) | |
<500 cm/Hoμr | |
7.87 in. | |
Certificate of Analysis | |
Highly Cross-Linked Agarose | |
20% Ethanol |
Stable in aqueous Buffers commonly used in Protein A Chromatography | |
2°C to 8°C | |
MabSelect SuRe Media | |
85 μm | |
20 cm | |
<200 kPa | |
3 to 12 | |
Alkali-stabilized Protein A-derived (E. coli) |
Safety and Handling
- MabSelect SuRe, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.