Learn More
Gibco™ DPBS, no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14190342
299.25 GBP valid until 2025-12-31
Use promo code "24341" to get your promotional price.
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
No Sodium Pyruvate | |
Storage conditions: 15°C to 30°C Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Gibco™ | |
5 L | |
Mammalian Cell Culture | |
7.0 to 7.3 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
1 X | |
DPBS | |
Dulbecco's Phosphate Buffered Saline | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3023175 | Certificate of Analysis | 13/02/2025 | 14190359,14190342,14190144,14190136,14190367 |
3042543 | Certificate of Analysis | 12/02/2025 | 14190359,14190342,14190144,14190136,14190367 |
3042529 | Certificate of Analysis | 12/02/2025 | 14190359,14190342,14190144,14190136,14190367 |
3062756 | Certificate of Analysis | 12/02/2025 | 14190359,14190342,14190144,14190136,14190367 |
2997815 | Certificate of Analysis | 12/02/2025 | 14190359,14190342,14190144,14190136,14190367 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.