missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MacConkey Agar without Salt
Detect and enumerate coliforms in urine samples to diagnose UTIs on this differential medium which prevents Proteus swarming.
£23.10
Specifications
Color | Salmon pink, transparent |
---|---|
Description | MacConkey Agar without Salt |
Format | 90mm Monoplate |
Manufacturing Origin | Germany |
Product Type | Agar |
Product Code | Brand | Fill Volume | Fill Weight | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fill Volume | Fill Weight | Price | Quantity & Availability | ||||
12972578
|
Thermo Scientific™
PO0148A |
19 mL | 19g ± 5% |
£23.10
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
Detect and enumerate coliform organisms in clinical samples with Thermo Scientific™ MacConkey Agar without Salt. The differential medium suppresses swarming of Proteus species and is, therefore, recommended for urine examination in the investigation of suspected urinary tract infection (UTI).
MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.Specifications
Salmon pink, transparent | |
90mm Monoplate | |
Agar | |
10/Pk. |
MacConkey Agar without Salt | |
Germany | |
Flow Wrapped - Single Layer |
Safety and Handling
missing translation for 'shelfLife' : 9 Weeks
For In Vitro Diagnostic Use.