missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma Macro Cuvette, 110-QX
Highly pure quartz glass cuvette with a PTFE stopper.
Brand: Hellma 110-5-46
Description
- High-purity quartz construction
- UV/VIS/NIR range
- PTFE stopper included
Specifications
1750 μL | |
PTFE | |
Base Thickness: 1.5 mm | |
1/Pk | |
200 to 3500 nm | |
12.5 mm |
Quartz Glass | |
9.5 mm | |
PTFE Stopper | |
5 mm | |
46 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction