Learn More
MP BIOMEDICALS MagBeads FastRNA Kit for FFPE Ready-to-Use for MPure-32 96 Preps
MagBeads FastRNA Kit for FFPE is intended for rapid extraction of RNA from tissue, cells, blood, and other clinical samples using the MPure-32%™ aNAP System. RNA can be used directly for RT-PCR, quantitative RT-PCR and so on.
Brand: MP BIOMEDICALS 117033500
Description
MagBeads FastRNA Kit for FFPE is based on the purification method of high binding magnetic particles. The sample is lysed and digested. RNA is released into the lysate.
After addition of magnetic particles and binding solution:
- RNA will be adsorbed on the surface of magnetic particles
- Impurities such as proteins will be removed without adsorption.
The adsorbed particles were washed with:
- Washing buffer to remove the proteins and impurities
- Fthanol to remove salts
Finally the RNA was eluted with Elution Buffer.
Specifications
Tissue, Cells, Blood, and Other Clinical Samples | |
RNA Extraction | |
RNA Kit for FFPE |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- (MPure-32) MagBeads FastRNA Kit for FFPE
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Dodecyl Sulphate, n-Hexadecane, Proteinase K, maleicacid, Ethanol, Guanidine Hydrochloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.