missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MagCellect Plus Human E-Cadherin+ Cell Isolation Kit

The MagCellect™ Plus Human E-Cadherin+ Cell Isolation Kit is designed to isolate Epithelial Cadherin (E-Cadherin) expressing cells via a positive selection principle. The resulting cell preparation is highly enriched with E-Cadherin+ cells. Typical purity of recovered E-Cadherin+ cells ranges from 80-95%.

Brand:  R&D Systems™ MAGH123

Product Code. 30002139

  • £788.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A mononuclear cell suspension is first incubated with the Human E-Cadherin Biotinylated Antibody that targets the desired cells. MagCellect™ Streptavidin (SA) Ferrofluid is next added to the reaction that allows the SA coated nanoparticles to interact with the cells tagged with the monoclonal antibodies. The tube containing the cell suspension is then placed within a magnetic field. Magnetically tagged cells will migrate toward the magnet (desired cell fraction), leaving the untagged cells (unwanted cell population) in suspension to be harvested by aspiration while the tube remains in the magnetic field. The tube containing the magnetically selected (desired) cells is then removed from the magnet and the cells are resuspended in MagCellect™ Plus Buffer or tissue culture media. The enriched cell preparation is then available for a variety of applications including tissue culture, immune status monitoring and flow cytometry

Kit Contents:

  • Human E-Cadherin Biotinylated Antibody (625 μL of Human E-Cadherin biotinylated antibody in a phosphate buffered solution containing BSA and preservative)
  • Human E-Cadherin Alexo Fluor™ 647 Antibody (125 μL of Alexa Fluor™ 647-conjugated Goat anti-human E-Cadherin Detection antibody)
  • Streptavidin Ferrofluid (1.25 mL of a solution containing BSA and preservatives)
  • Plus 2 bottles (50 mL/bottle) of a 10X concentrated buffer
Specifications

Specifications

999
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers