Learn More
Heathrow Scientific MagFuge™ High Speed Centrifuge Rotor
MagFuge™ High Speed Centrifuge Rotor for 12 x 1.5/2 mL tubes or 6 x 5 mL tubes.
Brand:  Heathrow Scientific 120622
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 16380886
  • £64.15 / Each
Estimated Shipment: 07-10-2024
to see stock.
Description
Description
  • High Speed Rotor
  • MagFuge Accessory
Specifications
Specifications
High Speed Centrifuge Rotor
12 x 1.5/2 mL
Centrifuge
12
GFPP
9,785 x g
Angled, Fixed