Learn More
Heathrow Scientific MagFuge™ High Speed Centrifuge Rotor
MagFuge™ High Speed Centrifuge Rotor for 12 x 1.5/2 mL tubes or 6 x 5 mL tubes.
Brand: Heathrow Scientific 120623
Additional Details : Weight : 0.12000kg
Description
- High Speed Rotor
- MagFuge Accessory
Specifications
High Speed Centrifuge Rotor | |
6 x 5 mL | |
Centrifuge | |
12 |
GFPP | |
9,785 x g | |
Angled, Fixed |