Learn More
Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
Brand:  Applied Biosystems™ A45469
79.05 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 16584810
  • £93.00 / Each
Explore more special offers
Description
Description
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60 mL Cell and Tissue DNA Extraction Buffer
Next-Generation Sequencing, PCR, Real-Time PCR
100 Preps
Cell and Tissue DNA Extraction Buffer
Room Temperature
Cells, Tissue
Automated Protocols, High-throughput Compatible, Manual Protocols
MagMAX™
100 Preps
Genomic DNA