Learn More
Applied Biosystems™ MagMAX™ Dx Viral/Pathogen NA Isolation Kit
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.
Brand: Applied Biosystems™ A52735
3113.12 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from respiratory biological specimens for downstream qPCR applications.
Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• Compatible with multiple downstream assays
• Single purification protocol for routinely tested pathogens and respiratory biological specimen without need for additional lysis
• Ability to purify various samples on single plate
• Sustainable product with biodegradable ingredients
• Designed to work with KingFisher Apex Dx instrument in under 40 mins
The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic-bead purification and in vitro diagnostic procedures.
Specifications
• Binding Solution, 530 mL |
|
Magnetic Bead | |
KingFisher™ Apex Dx | |
Up to 2,000 preps | |
MagMAX™ | |
Respiratory Pathogens |
96-well Plate | |
Nasopharyngeal swab, Anterior nasal swab, Saliva, Sputum | |
Real-Time PCR | |
Viral and Bacterial | |
Room Temperature |
For In Vitro Diagnostic Use