Learn More
Applied Biosytstems™ MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi.
Brand: Applied Biosytstems™ A59053
Additional Details : CAS Number : 12650-88-3 Weight : 0.00600kg
Description
MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.
Specifications
Magnetic | |
Store at –15 to –25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
DNA/RNA | |
Reagent | |
Dry Ice |
Clear | |
Bottle | |
Swabs in Transport Media, Saliva, Urine, Serum, Plasma | |
High-throughput Compatible, Automated Protocols | |
Up to 100 Reactions | |
MagMAX™ | |
200–400 μL | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |