missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosytstems™ MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi.
£244.00 - £2443.00
Specifications
Bead Type | Magnetic |
---|---|
Color | Clear |
Content And Storage | Store at –15 to –25°C. |
Format | Bottle |
Isolation Technology | Beads |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
17924741
|
Applied Biosytstems™
A59053 |
Up to 100 Reactions |
£244.00
5mL |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17914731
|
Applied Biosytstems™
A58153 |
Up to 1000 Reactions |
£2443.00
50mL |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.
Specifications
Magnetic | |
Store at –15 to –25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
MagMAX™ | |
200–400 μL | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |
Clear | |
Bottle | |
Swabs in Transport Media, Saliva, Urine, Serum, Plasma | |
High-throughput Compatible, Automated Protocols | |
DNA/RNA | |
Reagent | |
Dry Ice |