missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Aluminum Zinc rod, 9.53mm (0.375in) dia, 90cm (35in) long, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 014089.RM
View more versions of this product
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium Aluminum Zinc rod | |
Mg:Al:Zn; 96:3:1 wt% | |
90 cm | |
≈111.7g/90cm |
9.53mm (0.375 in.) dia., 90cm (35 in.) long | |
MFCD01074901 | |
Odorless | |
Rod |
RUO – Research Use Only