missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Aluminum Zinc rod, 9.53mm (0.375in) dia, 90cm (35in) long, Thermo Scientific Chemicals

£67.30 - £246.00

Specifications

Chemical Name or Material Magnesium Aluminum Zinc rod
Name Note 9.53mm (0.375 in.) dia., 90cm (35 in.) long
Avg. Mol. Wt. or Mol. Wt. Range Mg:Al:Zn; 96:3:1 wt%
MDL Number MFCD01074901
Odor Odorless
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014089.RM
90 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11307697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014089.RN
5 x 90 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium Aluminum Zinc rod
Mg:Al:Zn; 96:3:1 wt%
Odorless
Rod
9.53mm (0.375 in.) dia., 90cm (35 in.) long
MFCD01074901
≈111.7g/90cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only