missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 014092.RN
View more versions of this product
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Magnesium Aluminum Zinc wire | |
Mg:Al:Zn; 93:6:1 wt% | |
5 x 90 cm | |
Odorless | |
Wire |
3.18mm (0.125 in.) dia., 90cm (35 in.) long | |
MFCD01074901 | |
Insoluble in water. | |
≈12.44g/90cm |