Learn More
Magnesium chloride, pure, Thermo Scientific Chemicals
£27.84 - £500.49
Chemical Identifiers
CAS | 7786-30-3 |
---|---|
Molecular Formula | Cl2Mg |
Molecular Weight (g/mol) | 95.21 |
MDL Number | MFCD00011106 |
InChI Key | TWRXJAOTZQYOKJ-UHFFFAOYSA-L |
Synonym | magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride |
PubChem CID | 5360315 |
ChEBI | CHEBI:6636 |
SMILES | [Mg++].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697473
|
Thermo Scientific Acros
223211000 |
100 g | Plastic bottle |
£27.84
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10159872
|
Thermo Scientific Acros
223210010 |
1 kg | Plastic bottle |
£45.73
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10742685
|
Thermo Scientific Acros
223210025 |
2.5 kg | Plastic bottle |
£91.56
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498478
|
Thermo Scientific Acros
223210250 |
25 kg | Fibre drum |
£500.49
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7786-30-3 | |
95.21 | |
TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
5360315 | |
[Mg++].[Cl-].[Cl-] |
Cl2Mg | |
MFCD00011106 | |
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
CHEBI:6636 |
Specifications
1412.0°C | |
7786-30-3 | |
Powder | |
MgCl2 | |
11,31 | |
magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride | |
TWRXJAOTZQYOKJ-UHFFFAOYSA-L | |
magnesium;dichloride | |
5360315 | |
95.21 | |
Pure | |
Magnesium chloride |
708.0°C | |
White | |
Cl2Mg | |
MFCD00011106 | |
15,5726 | |
Solubility in water: 540g/L (20°C) | |
[Mg++].[Cl-].[Cl-] | |
95.21 | |
CHEBI:6636 | |
≥98.0% (on dry substance) | |
1.5% max. (105°C, 2 Hrs) |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 232-094-6
- Magnesium chloride, anhydrous
RUO – Research Use Only