Learn More
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
£37.20 - £512.00
Chemical Identifiers
CAS | 7439-95-4 |
---|---|
Molecular Formula | Mg |
Molecular Weight (g/mol) | 24.30 |
MDL Number | MFCD00085308 |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
Synonym | magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs |
PubChem CID | 5462224 |
ChEBI | CHEBI:25107 |
IUPAC Name | magnesium |
SMILES | [Mg] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391288
|
Thermo Scientific Alfa Aesar
010233.09 |
10 g |
£37.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301298
|
Thermo Scientific Alfa Aesar
010233.22 |
100 g |
£65.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311298
|
Thermo Scientific Alfa Aesar
010233.36 |
500 g |
£165.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381288
|
Thermo Scientific Alfa Aesar
010233.57 |
4 x 500 g |
£512.00
2000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
5462224 | |
magnesium |
Mg | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
CHEBI:25107 | |
[Mg] |
Specifications
7439-95-4 | |
Mg | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
-325 Mesh | |
Magnesium powder |
99.8% | |
MFCD00085308 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
24.3 | |
Powder |
Safety and Handling
P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
H250-H260-H500
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: II
missing translation for 'einecsNumber' : 231-104-6
missing translation for 'rtecsNumber' : OM2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Magnesium powder
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H228-Flammable solid.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
- P223-Do not allow contact with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P422-Store contents under inert gas.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only