missing translation for 'onlineSavingsMsg'
Learn More
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals Product Code.: 11391288

Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals

Product Code. 11391288
10 g
Click to view available options
Quantity:
10 g
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11391288

Brand: Thermo Scientific Alfa Aesar 010233.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Assay Percent Range 99.8%
Quantity 10 g
Formula Weight 24.3
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Magnesium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN05L022 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog Number010233.09,010233.22,010233.36,010233.57
Lot NumberX09K011 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number010233.09,010233.22,010233.36,010233.57
Lot NumberY30J024 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number010233.09,010233.22,010233.36,010233.57
Lot NumberH24W002 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010233.09,010233.22,010233.36,010233.57
Lot NumberI13Q16 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010233.09,010233.22,010233.36,010233.57
compliance-icons
Product Identifier
  • Magnesium powder
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P422-Store contents under inert gas.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.