Learn More
Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads
Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.
Brand: Invitrogen™ MSPB-6003-71
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Description
- Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
- The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
- For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
Specifications
25 Tests | |
Streptavidin | |
Biotin |
Liquid | |
Streptavidin Conjugate (Fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.