missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads
Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.
£69.50 - £256.00
Specifications
Form | Liquid |
---|---|
Ligand | Streptavidin |
Product Type | Streptavidin Conjugate (Fluorescent) |
Target Molecule | Biotin |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15592567
|
Invitrogen™
MSPB-6003-74 |
200 Tests |
£256.00
200 tests |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
15582567
|
Invitrogen™
MSPB-6003-71 |
25 Tests |
£69.50
25 tests |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
- The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
- For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
Specifications
Liquid | |
Streptavidin | |
Streptavidin Conjugate (Fluorescent) | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.