Learn More
Medline Scientific™ Magnum Trinocular Microscope
The trinocular version of the Magnum series with a choice of objectives types and LED illumination if preferred.
Brand:  Medline Scientific™ 2738.0000M
 View more versions of this product
Product Code. 15592947
  • £3020.00 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Trinocular optical head, inclined at 30°
  • 10x/20mm wide-field eyepiece
  • Available with either semi-planachromatic, planachromatic or phase contrast objectives
  • Brightfield Abbe condenser, NA 0.90 – NA 1.25 (oil)
Specifications
Specifications
Phase contrast 5-position turret
WF10X/20 mm
Halogen
Trinocular Biological Microscope
Quintuple reversed and revolving with click stop
220/240 V
Iris
Trinocular, Optical, with Vertical Photo/Video Port
Phase Contrast
Blue and green filter, 1 spare bulb, 2 spare fuses, immersion oil 10mL, soft rubber eyecups, dust cover and instruction manual.
220/240 V 50/60 Hz
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products