Learn More
Novex™ MAGPIX™ Performance Verification Kit
MAGPIX™ Performance Verification Kit
Brand: Novex™ MPXPVERK25
472.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
The convenient performance verification kit contains xMAP™ verification reagents that are used to verify calibration of the MAGPIX™ classification and reporter channels and fluidic integrity. Verification uses system controls to help ensure that the analyzer is functioning properly with current calibration settings. The kit is intended to be used with the off-plate reagent block. Each kit provides 25 verifications.Each kit includes:
Stripwells
The MAGPIX™ system is a versatile, compact, fluorescent detection system based on Luminex's proven xMAP™ technology. The system is a simple immunoassay platform designed for the simultaneous analysis of up to 50 intracellular and extracellular markers in single biological samples. The MAGPIX™ system provides users with a powerful multiplexing tool that offers higher sample throughput, increased assay flexibility, reduced sample volume, and lower costs than traditional ELISA and Western blotting.
For research use only. Not intended for human or animal therapeutic or diagnostic use.

Specifications
25 reactions | |
MAGPIX™ | |
Wet Ice | |
Luminex Instruments, MAGPIX System |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B92571 | Certificate of Analysis | 29/01/2025 | MPXPVERK25 |
B93966 | Certificate of Analysis | 29/01/2025 | MPXPVERK25 |
B95254 | Certificate of Analysis | 29/01/2025 | MPXPVERK25 |
B96515 | Certificate of Analysis | 29/01/2025 | MPXPVERK25 |
B98826 | Certificate of Analysis | 29/01/2025 | MPXPVERK25 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
27Hours
19Minutes
5Seconds
14 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
58Hours
19Minutes
5Seconds
14New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.