missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences MagStrep Strep-Tactin™ XT Beads
GREENER_CHOICE

MagStrep Strep-Tactin™ XT beads are magnetic beads (5% suspension) to purify Strep-tag™ II or Twin-Strep-tag™ fusion proteins.

£198.64 - £674.00

Specifications

Formulation 5% (v/v) Suspension in PBS with 0.1% (w/v) Sodium Azide
Support Type 6% Agarose, Crosslinked
Particle Size 20 to 40 μm
Form Resin Slurry
Target Twin-Strep-tag or Strep-tag II
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17942451
GREENER_CHOICE
View Documents
IBA Lifesciences
2-5090-002
2 mL
£198.64
2mL
Please to purchase this item. Need a web account? Register with us today!
17932451
GREENER_CHOICE
View Documents
IBA Lifesciences
2-5090-010
10 mL
£674.00
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.
  • Beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-Tactin™ XT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP).
  • MagStrep Strep-Tactin™ XT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.
  • The beads can be easily and swiftly separated with a magnetic separator. This eliminates the need for centrifugation and reduces the risk of disturbing the pellet or loss of beads within the wash step to zero.
  • The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.
Specifications

Specifications

5% (v/v) Suspension in PBS with 0.1% (w/v) Sodium Azide
20 to 40 μm
Twin-Strep-tag or Strep-tag II
6% Agarose, Crosslinked
Resin Slurry
Magnetic Agarose Bead
Videos
SDS
Documents

Documents

Product Certifications

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IBA Lifesciences MagStrep Strep-Tactin™ XT Beads

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.