Learn More
Emma Safety Footwear Mammoet M1807 XD Safety Shoes
Extra high boot is heat and cold insulated. Ankle protection Comfort Sponge with added padding provides a more comfortable feel and better fit. Mammoet M1807 is very easy to put on and take off; the vertical eyelets on the shoe make lacing the fire-retardant laces very simple.
Brand: Emma Safety Footwear 765868-43
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Heat and cold insulated
- Fire-retardant laces
- Waterproof Oil Nubuck construction
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver / Sympatex™ | |
43 |
Laces | |
Mammoet M1807 XD | |
Oiled Nubuck | |
Steel |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Emma Safety Footwear Mammoet M1807 XD Safety Shoes > Size: 43
Spot an opportunity for improvement?Share a Content Correction