Learn More
Manganese(II) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
£109.00 - £400.00
Chemical Identifiers
CAS | 1344-43-0 |
---|---|
Molecular Formula | MnO |
Molecular Weight (g/mol) | 70.94 |
MDL Number | MFCD00016226 |
InChI Key | VASIZKWUTCETSD-UHFFFAOYSA-N |
Synonym | manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide |
PubChem CID | 14940 |
IUPAC Name | oxomanganese |
SMILES | O=[Mn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460740
|
Thermo Scientific Alfa Aesar
044441.09 |
10 g |
£109.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470740
|
Thermo Scientific Alfa Aesar
044441.18 |
50 g |
£400.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-43-0 | |
70.94 | |
VASIZKWUTCETSD-UHFFFAOYSA-N | |
14940 | |
O=[Mn] |
MnO | |
MFCD00016226 | |
manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide | |
oxomanganese |
Specifications
1344-43-0 | |
Chunks and/or Powder | |
MFCD00016226 | |
Soluble in hydrochloric acid. Insoluble in water. | |
O=[Mn] | |
70.94 | |
70.94 | |
Odorless | |
(metals basis) |
Green | |
MnO | |
manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide | |
VASIZKWUTCETSD-UHFFFAOYSA-N | |
oxomanganese | |
14940 | |
99.99% | |
5.0 g/mL | |
Manganese (II) oxide |
Safety and Handling
GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
missing translation for 'einecsNumber' : 215-695-8
missing translation for 'rtecsNumber' : OP0900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only