Learn More
Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
£25.23 - £107.54
Chemical Identifiers
CAS | 1344-43-0 |
---|---|
Molecular Formula | MnO |
Molecular Weight (g/mol) | 70.94 |
MDL Number | MFCD00016226 |
InChI Key | VASIZKWUTCETSD-UHFFFAOYSA-N |
Synonym | manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxideShow More |
PubChem CID | 14940 |
IUPAC Name | oxomanganese |
SMILES | O=[Mn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11904151
|
Thermo Scientific Acros
436970010 |
1 g | Glass bottle |
£25.23
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914151
|
Thermo Scientific Acros
436970100 |
10 g | Glass bottle |
£107.54
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
1344-43-0 | |
70.94 | |
VASIZKWUTCETSD-UHFFFAOYSA-N | |
14940 | |
O=[Mn] |
MnO | |
MFCD00016226 | |
manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxideShow More | |
oxomanganese |
Specifications
1650.0°C | |
Green | |
76.7 to 78.2% (Mn) (Complexometry) | |
MFCD00016226 | |
Solubility in water: insoluble | |
O=[Mn] | |
70.94 | |
70.94 | |
Trace Metal Basis |
1344-43-0 | |
Crystalline Powder and/or Chunks | |
MnO | |
manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide | |
VASIZKWUTCETSD-UHFFFAOYSA-N | |
oxomanganese | |
14940 | |
99.99% | |
Manganese(II) oxide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 215-695-8
- Manganese(II) oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.