missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 11904151

Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 11904151
1 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11904151

Brand: Thermo Scientific Acros 436970010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-43-0
Molecular Formula MnO
Molecular Weight (g/mol) 70.94
MDL Number MFCD00016226
InChI Key VASIZKWUTCETSD-UHFFFAOYSA-N
Synonym manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxideShow MoreShow Less
PubChem CID 14940
IUPAC Name oxomanganese
SMILES O=[Mn]

Specifications

Melting Point 1650.0°C
Color Green
Physical Form Crystalline Powder and/or Chunks
Quantity 1 g
Assay Percent Range 76.7 to 78.2% (Mn) (Complexometry)
Solubility Information Solubility in water: insoluble
Formula Weight 70.94
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Manganese(II) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466313 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number436970010,436970100
Lot NumberA0426689 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number436970010,436970100
Lot NumberA0455695 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number436970010,436970100
Lot NumberA0449872 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number436970010,436970100
Lot NumberA0405977 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number436970010,436970100
Product Identifier
  • Manganese(II) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.