missing translation for 'onlineSavingsMsg'
Learn More
Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals Product Code.: 11317897

Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals

Product Code. 11317897
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11317897

Brand: Thermo Scientific Alfa Aesar 014340.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagentManganese(IV) oxide is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunsteinShow MoreShow Less
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Color Black
Physical Form Powder
Quantity 500 g
Merck Index 14,5730
Solubility Information Insoluble in water,nitric acid,sulfuric acid; Soluble in HCl; In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4
Formula Weight 86.94
Grade Technical
Concentration or Composition (by Analyte or Components) Mn 58% min.
Density 5.026 g/mL
Chemical Name or Material Manganese(IV) oxide, Activated
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP09J036 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number014340.09,014340.36,014340.22
Lot NumberP31J022 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number014340.09,014340.36,014340.22
Lot NumberZ23H035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014340.09,014340.36,014340.22
Lot NumberR31D022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014340.09,014340.36,014340.22
Lot NumberC01R008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014340.09,014340.36,014340.22
compliance-icons compliance-icons
Product Identifier
  • Manganese(IV) oxide, activated
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Manganese oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.