Learn More
Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals
Description
As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagentManganese(IV) oxide is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent

Specifications
Specifications
Melting Point | 535°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,5730 |
Solubility Information | Insoluble in water,nitric acid,sulfuric acid; Soluble in HCl; In presence of hydrogen peroxide or oxalic acid soluble in dilute HNO3 and H2SO4 |
Formula Weight | 86.94 |
Grade | Technical |
Concentration or Composition (by Analyte or Components) | Mn 58% min. |
Density | 5.026 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P09J036 | Certificate of Analysis | 09/05/2023 | 014340.09,014340.36,014340.22 |
P31J022 | Certificate of Analysis | 09/05/2023 | 014340.09,014340.36,014340.22 |
Z23H035 | Certificate of Analysis | 12/04/2022 | 014340.09,014340.36,014340.22 |
R31D022 | Certificate of Analysis | 08/12/2021 | 014340.09,014340.36,014340.22 |
C01R008 | Certificate of Analysis | 08/12/2021 | 014340.09,014340.36,014340.22 |
Safety and Handling


- Manganese(IV) oxide, activated
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Manganese oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.