Learn More
Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals
Crimping tools provide a reproducible, secure vial closure.
Brand:  Fisherbrand™ 13 06 0069
374.23 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11777597
  • £428.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13mm Crimper
Steel
For Flip Top/for Flip Off
13 mm
Not Autoclavable